missing translation for 'onlineSavingsMsg'
Learn More

Westfield Medical Sterilisation bag plain closure medical grade paper 125mm x 50mm x 250mm

Brand:  Westfield Medical RPC

Sterilisation bag plain closure medical grade paper 125mm x 50mm x 250mm

Code : 03

Additional Details : Weight : 0.20000kg

Product Code. 12769095

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.