Learn More
Sterilization Bags with Heat Seal Closure
Brand:  Westfield Medical MHS
Code : 03
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 12729115
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
CE Marked
Suitable For Steam and ETO Sterilisation, amd formaldehyde
250/pk
75mm
Heat seal
530mm
300mm
Videos
Alternative Products