Sterilin™ UTM (Universal Transport Medium) Swabs
Simplify collection, transport, maintenance and frozen storage of virus, chlamydia, mycoplasma and ureaplasma specimens. Available as filled tube with separate swab or in a kit.
Overview
Brand: Thermo Scientific Sterilin
Manufacturer Part Number: 359C
UNSPSC: 42141500
Code: Z3
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 12874641
Quantity Price
1 £ 138.0 / Pack of 50
In Stock 4
Add to basket
Description and Specification
Specification
Sterile AS/IRR
No. per Case 50
Applicator Material Plastic
Certifications/Compliance CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
Description 1mL medium, regular flocked swab
Tip Material Nylon