missing translation for 'onlineSavingsMsg'
Learn More

Sterilin™ Silicone Tubing

Perform medical, pharmaceutical and food applications with high quality, nontoxic Thermo Scientific™ Sterilin™ Silicone Tubing.

Brand:  Thermo Scientific Sterilin TSR1270320P

Code : 04

Additional Details : Weight : 3.70000kg

 View more versions of this product

Product Code. 13131454

  • £281.00 / 15m


In Stock 1
Add to basket

Description

Description

  • Laser etched batch code for ease of traceability
  • Ideal for use with peristaltic pumps
  • Can be autoclaved or dry heat sterilized
  • 15m length
  • Aseptically manufactured
Specifications

Specifications

Silicone tubing
19.1mm
49 ft.
3.2mm
12.7mm
Silicone
15m
Videos