Learn More
Thermo Scientific™ Sterilin™ Plain Swabs
Simplify sample collection with Thermo Scientific™ Sterilin™ Plain Swabs. Ideal for samples that will be processed immediately and don't require preservative for transport.
£295.92
Specifications
Certifications/Compliance | CE Marked: Class IIa, Medical Device Directive 93/42/EEC |
---|
Product Code | Brand | For Use With (Application) | Applicator Material | Packaging | No. per Case | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Applicator Material | Packaging | No. per Case | Price | Quantity & Availability | ||
11329633
|
Thermo Scientific™
165KS100 |
General purpose | Wood | Peel pouch | 2,500 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Simplify sample collection with Thermo Scientific™ Sterilin™ Plain Swabs. Ideal for samples that will be processed immediately and don't require preservative for transport. Supplied in either a peel pouch or in a tube for ease of handling and transportation.
AVAILABLE IN UK ONLY
- Primary sample collection device for bacteria
- Color-coded caps for ease of identification
Certifications
- CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
Notes
- Must be processed as soon as possible after sampling (ideally within two-hour period) to avoid dehydration of the sample and possible nonrecovery of pathogens
Ordering Information
- Not available in all geographic regions
Specifications
CE Marked: Class IIa, Medical Device Directive 93/42/EEC |