Learn More
Sterilin™ Plain Swabs
Simplify sample collection with Thermo Scientific™ Sterilin™ Plain Swabs. Ideal for samples that will be processed immediately and don't require preservative for transport.
Brand:  Thermo Scientific Sterilin F152CA
Code : Z4
Additional Details : Weight : 0.60000kg
 View more versions of this product
Product Code. 13171284
  • £34.35 / Pack of 100
Estimated Shipment
01-06-2018
Add to basket
Specifications
Specifications
Tube
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
IRR
1
Black
Wood
Charcoal neutralizes inhibitory effects of cotton
Cotton, Charcoal coated
100
Videos