Learn More
Sterilin™ Plain Swabs
Simplify sample collection with Thermo Scientific™ Sterilin™ Plain Swabs. Ideal for samples that will be processed immediately and don't require preservative for transport.
Brand:  Thermo Scientific Sterilin F150CA
Code : 56
Additional Details : Weight : 0.52000kg
 View more versions of this product
Product Code. 11349173
  • £38.45 / Pack of 100
In Stock 41
Add to basket
Specifications
Specifications
Tube
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
IRR
1
Red
Wood
General purpose
Cotton
100
Videos
Documentation