Learn More
Sterilin™ Plain Swabs
Simplify sample collection with Thermo Scientific™ Sterilin™ Plain Swabs. Ideal for samples that will be processed immediately and don't require preservative for transport.
Brand:  Thermo Scientific Sterilin F160CA
Code : 56
Additional Details : Weight : 5.38000kg
 View more versions of this product
Product Code. 11319293
  • £109.00 / Pack of 100
Estimated Shipment
22-02-2018
Add to basket
Specifications
Specifications
Tube
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
IRR
1
Orange
Aluminium
Ear, nose, vaginal or urethral
Rayon
100
Videos
Documentation
Technical Documents