Sterilin™ Plain Swabs
Simplify sample collection with Thermo Scientific™ Sterilin™ Plain Swabs. Ideal for samples that will be processed immediately and don't require preservative for transport.
Overview
Brand: Thermo Scientific Sterilin
Manufacturer Part Number: 451CST
X100 plain swab plastic/cotton vegetal protein coated
UNSPSC: 42141500
Code: Z4
Additional Details:
Additional Details: Weight: 0.60000kg
Product Code. 13147743
Quantity Price
1 £ 30.1 / Pack of 100
Estimated Shipment
27-02-2017
Add to basket
Description and Specification
Specification
Sterile IRR
Tip Material Cotton, Vegetal protein coated
No. per Pack 1
No. per Case 100
Cap Color Yellow
Packaging Tube
Applicator Material Plastic
Certifications/Compliance CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
For Use With (Application) Vegetal protein coating reduces overgrowth