Learn More
Sterilin™ Plain Swabs
Simplify sample collection with Thermo Scientific™ Sterilin™ Plain Swabs. Ideal for samples that will be processed immediately and don't require preservative for transport.
Brand:  Thermo Scientific Sterilin 165KS01
Code : 56
Additional Details : Weight : 1.76000kg
 View more versions of this product
Product Code. 11319633
  • £228.00 / Pack of 1000
In Stock 5
Add to basket
Specifications
Specifications
Peel pouch
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
IRR
1
N/A
Wood
General purpose
Cotton
1,000
Videos
Documentation
Technical Documents