missing translation for 'onlineSavingsMsg'
Learn More

Sterilin™ Plain Flocked Swabs

Optimize your samples with our innovative nylon fiber technology flocked swabs. Thermo Scientific™ Sterilin™ Plain Flocked Swabs provide both outstanding collection and up to 95% release.

Brand:  Thermo Scientific Sterilin 551C

Code : 04

Additional Details : Weight : 0.52500kg

 View more versions of this product

Product Code. 11389173

  • £164.00 / Pack of 100


Estimated Shipment
17-05-2018
Add to basket

Specifications

Specifications

Minitip nylon flocked swab
Plastic
EO
100
Flocked swabs - Individually wrapped in plain dry tube
Individually wrapped in plain dry tube
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use); Certified Human DNA-free, RNase/DNase-free
Nylon
80mm
Videos
Documentation