Learn More
Sterilin™ Gel Transport Swabs
Ensure optimal sample collection and recovery using Thermo Scientific™ Sterilin™ Gel Transport Swabs. Swabs contain Amies media to preserve sample during lab transport.
Brand:  Thermo Scientific Sterilin 414CST
Code : 56
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 12862813
  • £309.00 / Pack of 500
Estimated Shipment
05-06-2018
Add to basket
Specifications
Specifications
M40 transport swab
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
IRR
50
Black
Plastic
Urogenital
Rayon
500
Amies w/charcoal
Videos
Documentation
Technical Documents