Learn More
Sterilin™ Gel Transport Swabs
Ensure optimal sample collection and recovery using Thermo Scientific™ Sterilin™ Gel Transport Swabs. Swabs contain Amies media to preserve sample during lab transport.
Brand:  Thermo Scientific Sterilin 18192CST
UNSPSC : 42141500
Code : 56
Additional Details : Weight : 8.62000kg
 View more versions of this product
Product Code. 11329173
  • £1110.00 / Pack of 500
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications Description & Specifications
Specifications
Standard transport swab
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
IRR
50
Blue
Twisted wire
Pernasal
Rayon
500
Amies w/charcoal
Documentation