Learn More
Sterilin™ Gel Transport Swabs
Ensure optimal sample collection and recovery using Thermo Scientific™ Sterilin™ Gel Transport Swabs. Swabs contain Amies media to preserve sample during lab transport.
Brand:  Thermo Scientific Sterilin 18190CST
Code : 56
Additional Details : Weight : 8.81500kg
 View more versions of this product
Product Code. 11319173
  • £1175.00 / Pack of 500
Estimated Shipment
26-01-2018
Add to basket
Specifications
Specifications
Standard transport swab
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
IRR
50
Blue
Twisted wire
Pernasal
Rayon
500
Amies
Videos
Documentation