Sterilin™ Gel Transport Swabs
Ensure optimal sample collection and recovery using Thermo Scientific™ Sterilin™ Gel Transport Swabs. Swabs contain Amies media to preserve sample during lab transport.
Overview
Brand: Thermo Scientific Sterilin
Manufacturer Part Number: 18116CST
UNSPSC: 42141500
Code: 56
Additional Details:
Additional Details: Weight: 7.48000kg
Product Code. 11309173
Quantity Price
1 £ 970.0 / Pack of 500
Estimated Shipment
08-06-2017
Add to basket
Description and Specification
Specification
Description Standard transport swab
Applicator Material Aluminium
Certifications/Compliance CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
For Use With (Application) Urogenital
Sterile IRR
Tip Material Rayon
No. per Pack 50
No. per Case 500
Cap Color Orange
Media Type Amies w/charcoal
Documentation