Sterilin™ Gel Transport Swabs
Ensure optimal sample collection and recovery using Thermo Scientific™ Sterilin™ Gel Transport Swabs. Swabs contain Amies media to preserve sample during lab transport.
Overview
Brand: Thermo Scientific Sterilin
Manufacturer Part Number: 18110CST
UNSPSC: 42141500
Code: 56
Additional Details:
Additional Details: Weight: 7.61500kg
Product Code. 11399163
Quantity Price
1 £ 985.0 / Pack of 500
Estimated Shipment
16-05-2017
Add to basket
Description and Specification
Specification
Media Type Amies
Description Standard transport swab
No. per Case 500
Sterile IRR
Applicator Material Aluminium
Certifications/Compliance CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
Tip Material Rayon
Cap Color Orange
For Use With (Application) Paediatric or urethral
No. per Pack 50
Documentation