Learn More
Sterilin™ Gel Transport Swabs
Ensure optimal sample collection and recovery using Thermo Scientific™ Sterilin™ Gel Transport Swabs. Swabs contain Amies media to preserve sample during lab transport.
Brand:  Thermo Scientific Sterilin 18111CST
Code : 56
Additional Details : Weight : 7.54000kg
 View more versions of this product
Product Code. 11389163
  • £515.00 / Pack of 500
Estimated Shipment
12-02-2018
Add to basket
Specifications
Specifications
Standard transport swab
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
IRR
50
Blue
Plastic
General purpose
Rayon
500
Stuart agar
Videos
Documentation