Learn More
Sterilin™ Gel Transport Swabs
Ensure optimal sample collection and recovery using Thermo Scientific™ Sterilin™ Gel Transport Swabs. Swabs contain Amies media to preserve sample during lab transport.
Brand:  Thermo Scientific Sterilin 18108CST
UNSPSC : 42141500
Code : 56
Additional Details : Weight : 7.80000kg
 View more versions of this product
Product Code. 11369163
  • £477.00 / Pack of 500
In Stock 6
Add to basket
Specifications Description & Specifications
Specifications
Standard transport swab
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
IRR
50
Blue
Plastic
Throat, vagina, wound and skin
Rayon
500
Amies
Documentation