missing translation for 'onlineSavingsMsg'
Learn More

Sterilin™ Environmental Swabs

Monitor surface conditions in your facilities with our Thermo Scientific™ Sterilin™ Environmental Swabs. Swab and transport with our easy-to-use kits.

Brand:  Thermo Scientific Sterilin 905C

Code : 56

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12802833

  • £148.00 / Pack of 10


Estimated Shipment
05-02-2018
Add to basket

Specifications

Specifications

Square sampling kit
PS
IRR
10
10mL solution, separate swab, dacron tip with square sampling template
13cm
1
Videos
Documentation