missing translation for 'onlineSavingsMsg'
Learn More

Sterilin™ Environmental Swabs

Monitor surface conditions in your facilities with our Thermo Scientific™ Sterilin™ Environmental Swabs. Swab and transport with our easy-to-use kits.

Brand:  Thermo Scientific Sterilin 902C

Code : 56

Additional Details : Weight : 0.60000kg

 View more versions of this product

Product Code. 11389283

  • £78.00 / Pack of 25


Estimated Shipment
11-06-2018
Add to basket

Specifications

Specifications

Swab rinse kit
PS
IRR
25
10mL solution, separate swab, dacron tip
13cm
1
Videos
Documentation

Technical Documents