Learn More
Sterilin™ E-Swab and Transport System
Securely collect and transport clinical microbiological specimens. Pre-labeled container provided with 1mL liquid amies medium and a flocked swab.
Brand:  Thermo Scientific Sterilin 484CE
Code : 56
Additional Details : Weight : 0.60000kg
 View more versions of this product
Product Code. 12802863
  • £146.00 / Pack of 50
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
Transport swab, liquid amies, paediatric flocked swab
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
Nylon fiber
Plastic
Yes
50
Videos
Documentation
Technical Documents