Learn More
Sterilin™ E-Swab and Transport System
Securely collect and transport clinical microbiological specimens. Pre-labeled container provided with 1mL liquid amies medium and a flocked swab.
Brand:  Thermo Scientific Sterilin 482CE
Code : 56
Additional Details : Weight : 0.47000kg
 View more versions of this product
Product Code. 12882853
  • £157.00 / Pack of 50
In Stock 10
Add to basket
Specifications
Specifications
Transport swab, liquid amies, pernasal flocked swab
CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
Nylon fiber
Plastic
Yes
50
Videos
Documentation
Technical Documents