Learn More
BioTek™ Sterile Robotic System Pipette Tips
BioTek™ Sterile Robotic System Pipette Tips are suitable for use with the Precision™ Microplate Sample Processor and Pipetting System.
Brand:  BioTek™ 98193
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12575077
  • £956.00 / Pack of 50
Estimated Shipment
12-04-2021
Add to basket
Description
Description
  • Sterile
  • Suitable for Precision™ Microplate Sample Processor and Pipetting System
Specifications
Specifications
200 μL
Precision™ Microplate Sample Processor and Pipetting System
Rack
96 tips per SBS standard robotic rack. 10 racks per package. 5 packages per case.
Sterile
Pipette tips for robotic system
Automation
4,800 total tips