Learn More
BioTek™ Sterile Robotic System Pipette Tips
BioTek™ Sterile Robotic System Pipette Tips are suitable for use with the Precision™ Microplate Sample Processor and Pipetting System.
Brand: BioTek™ 98189
Additional Details : Weight : 0.80000kg
Description
- Sterile
- Suitable for Precision™ Microplate Sample Processor and Pipetting System
Specifications
50 μL | |
Precision™ Microplate Sample Processor and Pipetting System | |
Rack | |
96 tips per SBS standard robotic rack. 10 racks per package. 5 packages per case. |
Sterile | |
Pipette tips for robotic system | |
Automation | |
4,800 total tips |