Learn More
BioTek™ Sterile, Level Sense-Capable, Robotic System Pipette Tips
BioTek™ Sterile Level Sense-Capable Robotic System Pipette Tips are suitable for use with the Precision™ Microplate Sample Processor and Pipetting System.
Brand:  BioTek™ 98206
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12561307
  • £1427.00 / Pack of 4800
Estimated Shipment
06-04-2021
Add to basket
Description
Description
  • Sterile
  • Level sense-capable
  • Suitable for Precision™ XS Microplate Sample Processor and Pipetting System
Specifications
Specifications
50 μL
Precision™ XS Microplate Sample Processor and Pipetting System
Level sense capable
Automation
4800/pk
Sterile
Pipette tips for robotic system
Rack
96 tips per SBS standard robotic rack.  10 racks per package.  5 packages per case.