Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head

Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: 916-021

UNSPSC: 41121608

Code: 03

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Code. 12368562

Quantity Price
1 £ 133.0 / Pack of 960
Estimated Shipment
10-04-2017
Add to basket


Description and Specification

Specification

Color Clear
Volume (Metric) 40μL
Sterility Sterile
For Use With (Equipment) Beckman Liquid Handling Systems
Packaging 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Filtered Yes
Product Line ART

Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well pates
  • Low percentage coefficient of variation (%CV)