Learn More
Fisherbrand™ Sterile PES Syringe Filter
Designed for filtration and purification in a sterile environment. Fisherbrand™ Sterile PES Syringe Filter features a PES membrane in a polypropylene housing for purification of tissue culture solutions from a standard syringe. 33mm diameter membranes available with 0.2 or 0.45μm pore sizes.
Brand: Fisherbrand™
49.21 GBP valid until 2024-12-31
Use promo code "21934" to get your promotional price.
Code : NEW
Informations supplémentaires : Weight : 5.60250kg
Description
- Ideal for purification of tissue culture solutions and buffers
- High sample throughput with low protein binding
- Hydrophilic 33mm polyethersulfone (PES) membrane
- Sturdy, chemically resistant polypropylene housing
Spécification
White | |
0.45 μm | |
Purification of tissue culture solutions, buffers | |
50 Pack | |
33 mm |
PES | |
Sterile | |
Polypropylene | |
Syringe Filter |