missing translation for 'onlineSavingsMsg'
Learn More

Zeiss Microscope body DV4 240V 60Hz 100W 4x zoom stereo, for fitting into devices with 76mm

Brand:  Zeiss 4354219901000

Microscope body DV4 240V 60Hz 100W 4x zoom stereo, for fitting into devices with 76mm

Code : 04

Additional Details : Weight : 0.11100kg

Product Code. 10110240

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.