missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stella/Dppa3 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Stella/Dppa3 |
---|---|
Immunogen | E. coli-derived recombinant mouse Stella/Dppa3 Met1-Asn150 Accession # Q8QZY3 |
Host Species | Goat |
Primary or Secondary | Primary |
Classification | Polyclonal |
Description
Stella/Dppa3 Polyclonal specifically detects Stella/Dppa3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Stella/Dppa3 | |
Goat | |
Polyclonal | |
IgG | |
developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Western Blot, Immunohistochemistry | |
RUO |
E. coli-derived recombinant mouse Stella/Dppa3 Met1-Asn150 Accession # Q8QZY3 | |
Primary | |
Mouse | |
Unconjugated | |
359787 | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only