missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Steel Stand Bases
Steel Stand Bases with one adjustable foot.
Brand: Bochem™ 5038
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Thread M10
- 1 foot adjustable
Specifications
Steel | |
315 mm | |
Stand Base |
10 mm | |
200 mm | |
1 Ea. |