Learn More
Steel shot, non-spherical, 1mm (0.039in) dia, Thermo Scientific Chemicals
£62.90 - £126.00
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339758
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
044236.A3
2 kg
£62.90
2kg
View alternative products
Save Up to 
»
In Stock
to see stock.
11349758
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
044236.A7
5 kg
£126.00
5kg
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
Steel shot is used in cleaning applications for removal of loose material on metal surfaces. This type of cleaning is common in automotive industry (motor blocks, cylinder heads. Steel shot is also used in surface preparation process for cleaning metal surfaces which are covered with mill scale, dirt, rust, or paint coatings and for physically modifying the metal surface such as creating roughness for better application of paint and coating. The steel shots are generally employed in shot blasting machines.