missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Steel shot, non-spherical, 0.5mm (0.02 in.) dia.

Brand:  Alfa Aesar™ 044235.A3

Code : NEW

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 11319758

  • £32.90 / 2kg


EU Stock 1
Add to basket

Specifications

Specifications

Steel shot
0.5mm (0.02 in.) dia.
MFCD00678174
2kg
Non-Spherical
Videos
SDS