missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stearic Acid, Powder (Triple Pressed), N.F., J.T. Baker™
Click to view available options
Quantity:
500 g
1.5 kg
Packaging:
Poly Bottle
Specifications
Flash Point | 196.00°C |
Packaging | Poly Bottle |
Quantity | 500 g |
Specific Gravity | 1L = 0.94kg |
Formula Weight | 284.48 |
Grade | NF |
Physical Form | Powder (Triple Pressed) |
Chemical Name or Material | Stearic Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'