missing translation for 'onlineSavingsMsg'
Learn More

Stearic Acid, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073574
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
57-11-4
C18H36O2
284.48
MFCD00002752
QIQXTHQIDYTFRH-UHFFFAOYSA-N
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
5281
CHEBI:28842
octadecanoic acid
CCCCCCCCCCCCCCCCCC(O)=O
This item is not returnable. View return policy
57-11-4
C18H36O2
284.48
MFCD00002752
QIQXTHQIDYTFRH-UHFFFAOYSA-N
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
5281
CHEBI:28842
octadecanoic acid
CCCCCCCCCCCCCCCCCC(O)=O
Melting Point 67°C
Color White
Boiling Point 361°C
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 284.47g/mol
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'