missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Staurosporine, 98%
218.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 mg
250 μg
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Staurosporine is an indolocarbazole alkaloid capable of permeating cell membranes
• Staurosporine has anti-cancer properties that can induce apoptosis
• Staurosporine is a broad inhibitor of protein kinases, preventing growth and division cycles
Application
• Staurosporine is used for research on acute myeloid leukemia
• It is suitable for research applications including cell culture, in vivo studies, and biochemistry
Especificaciones
Melting Point | 270.0°C |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 250 μg |
Merck Index | 15,8928 |
Solubility Information | Solubility in water: .. Other solubilities: soluble in ethylacetate |
Formula Weight | 466.5 |
Percent Purity | 98% |
Chemical Name or Material | Staurosporine |
Seguridad y manipulación
Nombre del producto
- Staurosporine
- Warning
- Carcinogenicity Category 2
- H351-Suspected of causing cancer.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'