Learn More
Stattic, Tocris Bioscience™
Selective STAT3 inhibitor
Brand:  Tocris (Bio-Techne) 279850
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13461854
  • £278.53 / 50mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Small molecule inhibitor of STAT3. Inhibits binding of tyrosine-phosphorylated peptide motifs to STAT3 SH2 domain and inhibits STAT3 activation, dimerization and nuclear translocation. Displays selectivity over STAT1, STAT5, c-Myc/Max, Jun/Jun and Lck. Induces apoptosis in STAT3-dependent cancer cell lines.
Specifications
Chemical Identifiers
19983-44-9
211.191
stattic, 6-nitrobenzo b thiophene 1,1-dioxide, stat three inhibitory compound, stat3 inhibitor v, stattic, 6-nitro-1, stat3 inhibitor v, 6-nitrobenzo b thiophene-1,1-dioxide, 1,1-dioxide-6-nitro-benzo b thiophene, 6-nitro-1lambda6-benzothiophene-1,1-dione, benzo b thiophene, 6-nitro-, 1,1-dioxide
CHEBI:86989
C1=CC(=CC2=C1C=CS2(=O)=O)[N+](=O)[O-]
C8H5NO4S
ZRRGOUHITGRLBA-UHFFFAOYSA-N
2779853
6-nitro-1-benzothiophene 1,1-dioxide
Specifications
Stattic
19983-44-9
stattic, 6-nitrobenzo b thiophene 1,1-dioxide, stat three inhibitory compound, stat3 inhibitor v, stattic, 6-nitro-1, stat3 inhibitor v, 6-nitrobenzo b thiophene-1,1-dioxide, 1,1-dioxide-6-nitro-benzo b thiophene, 6-nitro-1lambda6-benzothiophene-1,1-dione, benzo b thiophene, 6-nitro-, 1,1-dioxide
C1=CC(=CC2=C1C=CS2(=O)=O)[N+](=O)[O-]
211.191
CHEBI:86989
>99%
50mg
C8H5NO4S
ZRRGOUHITGRLBA-UHFFFAOYSA-N
6-nitro-1-benzothiophene 1,1-dioxide
2779853
211.19
Videos