missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ STAT6 Recombinant Human Protein, His Tag

Suitable for use in protein studies such as protein structure analysis and protein-protein interactions

Brand:  Sino Biological™ 13190H08B250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15216235

  • £424.73 / 250µg


Estimated Shipment
08-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: 20mM Tris, 500mM NaCl, 20% Glycerol, 0.3mM DTT, pH 7.4
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
6778
≥85% by SDS-PAGE
Human
<1 EU/μg
STAT6
Other Proteins
STAT6 Recombinant Human Protein
Lyophilized
95.5kd
5 x 50μg
Full Length, Recombinant
Signal Transducer And Activator Of Transcription 6
Assay Standard
Insect
His Tag
Videos