Learn More
STARD9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553194
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GMYSEPLRQF RDSSVGDQNA QVCQTNPEPP ATTQGPHTLD LSEGSAESKL VVEPQHECLE NTTRCFLEKP QFSTELRDHN RLDSQAKFVA RLKHTCSPQE DSPWQEEEQH RDQASGGGE Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 52%.
STARD9 Polyclonal antibody specifically detects STARD9 in Human samples. It is validated for Immunohistochemistry.Specifications
STARD9 | |
Polyclonal | |
Unconjugated | |
STARD9 | |
KIAA1300; KIF16A; StAR related lipid transfer domain containing 9; StARD9; StAR-related lipid transfer (START) domain containing 9; StAR-related lipid transfer domain containing 9; stAR-related lipid transfer protein 9; START domain containing 9; START domain-containing protein 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57519 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2P6 | |
STARD9 | |
Recombinant protein corresponding to Human STARD9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only