missing translation for 'onlineSavingsMsg'
Learn More

Starch, Soluble, Technical, SLR, Fisher Chemical™

Brand:  Fisher Chemical S/7960/60

Code : 26

Additional Details : CAS Number : 9005-25-8 Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 10080410

  • £106.00 / 1kg


In stock, ready to ship: 32
Add to basket

Description

Description

Starch, Soluble, Technical, SLR

Specifications

Chemical Identifiers

9005-25-8
342.30
GUBGYTABKSRVRQ-UHFFFAOYNA-N
439341
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
C12H22O11
MFCD00082026,MFCD00132834
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
CHEBI:18167
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

9005-25-8
MFCD00082026,MFCD00132834
GUBGYTABKSRVRQ-UHFFFAOYNA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
439341
Powder Solid
White
1kg
C12H22O11
alpha-maltose, maltose, starch, soluble, glcalpha1-4glca, unii-15sug9ad26, glcalpha1-4glcalpha, amylodextrin, starch solution, alpha-malt sugar, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
342.30
CHEBI:18167
Plastic powder jar
5 to 7
Videos
SDS Documents
Certificates