missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Starch, Certified AR for Analysis, Soluble, Fisher Chemical™
79.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
250 g
500 g
1 kg
Description
C12H22O11, CAS Number-9005-25-8, starch, soluble, 4-o-alpha-d-glucopyranosyl-alpha-d-glucopyranose, glcalpha1-4glca, maltose, unii-15sug9ad26, starch solution, alpha-malt sugar, alpha-maltose, amylodextrin, glcalpha1-4glcalpha, 250g, CHEBI:18167, White, GUBGYTABKSRVRQ-ASMJPISFSA-N
Specifications
Color | White |
pH | 5 to 7 |
Physical Form | Powder Solid |
Quantity | 500 g |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'