missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stanniocalcin 2/STC-2 Rat anti-Human, Clone: 348217, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Stanniocalcin 2/STC-2 |
---|---|
Clone | 348217 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Stanniocalcin 2 Thr25-Arg302 Accession # O76061 |
Gene ID (Entrez) | 8614 |
Gene Accession No. | O76061 |
Description
Stanniocalcin 2/STC-2 Monoclonal specifically detects Stanniocalcin 2/STC-2 in Human samples. It is validated for Western Blot.Specifications
Stanniocalcin 2/STC-2 | |
Mouse myeloma cell line NS0-derived recombinant human Stanniocalcin 2 Thr25-Arg302 Accession # O76061 | |
O76061 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
348217 | |
8614 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human Stanniocalcin 2 (STC-2) in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) STC-1 or rmSTC-2. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only