Learn More
Fisher Chemical SPECTRAPURE - Standard (VI) 30 components - Ag, Al, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, K, Li , Mg, Mn, Mo, Na, Ni, Pb, Rb, Se, Sr, Te, Tl, U, V in HNO3 5%/tr. HF
Brand: Fisher Chemical PUR300125
SPECTRAPURE - Standard (VI) 30 components - Ag, Al, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, K, Li , Mg, Mn, Mo, Na, Ni, Pb, Rb, Se, Sr, Te, Tl, U, V in HNO3 5%/tr. HF
Code : 04
Additional Details : Weight : 0.12500kg Transport : UN number : 1760 Chem class : 8 Pack group : II