Learn More
Stainless Steel gauze, 20 mesh woven from 0.102mm (0.004in) dia wire, Type 316, Thermo Scientific Chemicals
Open area: 84.6%; Width of opening: 0.046 in.
Brand: Thermo Scientific Alfa Aesar 040943.HB
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel | |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
300 x 300 mm | |
20 Mesh |
Gauze | |
MFCD00148499 | |
Insoluble in water. | |
Wire Cloth |
RUO – Research Use Only