missing translation for 'onlineSavingsMsg'
Learn More

Stainless-Steel wire, annealed, Alfa Aesar™

Temper: soft

Brand:  Alfa Aesar 043301.BU

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 25cm

 View more versions of this product

Product Code. 11309448

  • £12.60 / 25cm


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

Stainless Steel
Wire, 0.38mm (0.015 in.) dia.
MFCD00148499
annealed, Type 304
Fe:Cr:Ni; 70:19:11 wt%

Specifications

Stainless Steel
25cm
Silver-colored wire
Wire, 0.38mm (0.015 in.) dia.
MFCD00148499
Silver
annealed, Type 304
Odorless
Fe:Cr:Ni; 70:19:11 wt%
Videos
SDS