missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Stainless Steel wire, 2.0mm (0.08 in.) dia., Type 304

Brand:  Thermo Scientific Alfa Aesar 040948.BW

Stainless Steel wire, 2.0mm (0.08 in.) dia., Type 304

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11335277

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.