missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Stainless Steel wire, 2.0mm (0.08 in.) dia., Type 304

Brand:  Alfa Aesar™ 040948.BZ

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 150cm

 View more versions of this product

Product Code. 11325277

  • £69.60 / 150cm


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

Stainless Steel wire, Type 304
2.0mm (0.08 in.) dia.
MFCD00148499

Specifications

Fe:Cr:Ni; 70:19:11 wt%
150cm
Odorless
MFCD00148499
Stainless Steel wire, Type 304
2.0mm (0.08 in.) dia.
Wire
Videos
SDS