missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Stainless Steel tubing, 1.65mm (0.065in) OD, 1.19mm (0.047in) ID, Type 304

Brand:  Thermo Scientific Alfa Aesar 013469.BV

Stainless Steel tubing, 1.65mm (0.065in) OD, 1.19mm (0.047in) ID, Type 304

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11366828

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.