missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Stainless Steel tubing, 0.82mm (0.032in) OD, 0.51mm (0.02in) ID, Type 304

Brand:  Alfa Aesar™ 013468.BV

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 30cm

 View more versions of this product

Product Code. 11346828

  • £14.00 / 30cm


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

Stainless Steel tubing, Type 304
OD: 0.82mm (0.032 in.), ID: 0.51mm (0.02 in.)
MFCD00148499

Specifications

Fe:Cr:Ni; 70:19:11 wt%
30cm
MFCD00148499
Stainless Steel tubing, Type 304
OD: 0.82mm (0.032 in.), ID: 0.51mm (0.02 in.)
Videos
SDS