missing translation for 'onlineSavingsMsg'
Learn More

Huber Kaeltmaschinenbau™ Stainless Steel Test Tube Rack

Brand:  Huber Kaeltmaschinenbau™ 6040

Code : NEW

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 10616717

  • £235.00 / Each


Estimated Shipment
18-06-2018
Add to basket

Specifications

Specifications

Stainless Steel
For 112A-E, 118A-E, 212B to 225B and K12 to K25
58
Videos