missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories for Analytical Filter Holders

Accessories for filter holders for sample preparation.

Brand:  Merck XF2004758

49.14 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : NEW

Additional Details : Weight : 3.50000kg

 View more versions of this product

Product Code. 10293871

  • £52.75 / Each
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

Accessories for filter holders for sample preparation.

  • Not Autoclaved
  • Specifications

    Specifications

    Not Autoclavable
    Non-sterile
    47 mm diameter
    Stainless Steel
    Support Frit
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers